missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ IL17F Recombinant Human Protein
Recombinant Human IL17F is a bioactive protein intended for use in cell culture applications
£69.50
Specifica
For Use With (Application) | Cell Culture |
---|---|
Molecular Weight (g/mol) | 38 kDa |
Purification Method | Gel-Purified |
Gene Alias | IL-17F |
Protein Length | 133 aa |
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
11390852
|
Gibco™
PHC9554 |
10μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Descrizione
IL17F is a member of structurally related cytokines. The IL-17 family of cytokines plays a role in mediating proinflamatory responses.- High purity: minimal interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to help ensure a biologically active protein
- High biological activity: in house activity testing includes a variety of cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by the dose-dependent induction of IL-6 secretion from NDHF Adult fibroblasts
Shipping Condition: Dry Ice.
Specifica
Cell Culture | |
Gel-Purified | |
133 aa | |
Recombinant | |
Carrier-Free | |
Induction of IL-6 secretion from NDHF cells | |
Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Neurobiology, Inflammation, Angiogenesis, Signal Transduction | |
90 % |
38 kDa | |
IL-17F | |
Human | |
Store in freezer (-5 to -30°C). | |
100 ng/ml | |
< 1 EU/ml | |
Mammalian | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.