missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL17F Recombinant Human Protein

Recombinant Human IL17F is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC9551

Code : 50

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 11370852

  • £626.24 / Each


Estimated Shipment
10-07-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

IL17F is a member of structurally related cytokines. The IL-17 family of cytokines plays a role in mediating proinflamatory responses.
  • High purity: minimal interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to help ensure a biologically active protein
  • High biological activity: in house activity testing includes a variety of cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by the dose-dependent induction of IL-6 secretion from NDHF Adult fibroblasts

Shipping Condition: Dry Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
Gel-Purified
100μg
Store at -5° to -30°C.
Recombinant/Ligand
<1 EU/mL
Induction of IL-6 secretion from NDHF cells
Cytokines & Receptors
133 aa
38kDa
90%
Human
Carrier Protein
Interleukins
IL-17F
Cell Culture
Mammalian
100ng/mL
Videos

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.