missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL17A Recombinant Human Protein

Recombinant IL17A is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC9174

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10768013

  • £214.50 / Each


Estimated Shipment
01-05-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

IL17A is a proinflammatory cytokine that regulates the activities of NFκB and mitogen-activated protein kinases. This cytokine can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO).

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
15.5kDa
95%
Human
Carrier Protein
Interleukins
IL-17A
Cytokines & Receptors
272 aa (homodimer)
Lyophilized
Sequential Chromatography
10μg
Store at -5° to -30°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Videos

For Research Use Only. Not for use in diagnostic procedures.