missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL17 Recombinant Mouse Protein

Recombinant IL17 is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMC0175

Code : 50

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10242533

  • £147.29 / Each


Estimated Shipment
08-06-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

IL17 stimulates epithelial, endothelial or fibroblastic cells to secrete IL-6, IL-8 and G-CSF. IL17 also increases the expression of ICAM-1 in fibroblasts.

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
16171
Sequential Chromatography
25μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
15kDa
95%
Mouse
Carrier Protein
Interleukins
IL-17
Cytokines & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.