missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL17 Recombinant Mouse Protein

Recombinant IL17 is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMC0174

Code : 50

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 10029472

  • £228.00 / Each

Estimated Shipment: 16-08-2021

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

IL17 stimulates epithelial, endothelial or fibroblastic cells to secrete IL-6, IL-8 and G-CSF. IL17 also increases the expression of ICAM-1 in fibroblasts.

Shipping Condition: Wet Ice.

Specifications

Specifications

< 0.1 ng/µg
Wet Ice
Lyophilized
Cytokines & Receptors
Interleukins
Immunology, Neurobiology, Inflammation, Angiogenesis, Signal Transduction
Cell Culture
15 kDa
95 %
IL-17
10 µg
E. coli
Gibco™
Recombinant, Ligand
Carrier-Free
Mouse
16171
Sequential Chromatography
10μg
Yes
Videos
SDS
Documents
Certificates

For Research Use Only. Not for use in diagnostic procedures.