missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL17 Recombinant Mouse Protein

Recombinant IL17 is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMC0171

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10019472

  • £514.00 / Each

Estimated Shipment
28-07-2020

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

IL17 stimulates epithelial, endothelial or fibroblastic cells to secrete IL-6, IL-8 and G-CSF. IL17 also increases the expression of ICAM-1 in fibroblasts.

Shipping Condition: Wet Ice.

Specifications

Specifications

Store in refrigerator (2–8°C).
100 µg
Carrier-Free
E. coli
Cytokines & Receptors
Interleukins
Mouse
16171
Sequential Chromatography
100μg
< 0.1 ng/µg
Gibco®
Wet Ice
Lyophilized
Recombinant, Ligand
Immunology, Neurobiology, Inflammation, Angiogenesis, Signal Transduction
Cell Culture
15 kDa
95 %
IL-17
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.