Learn More
Gibco™ Human IL-17A Recombinant Protein
Human IL-17A Recombinant Protein
Brand: Gibco™ PHC0174
Code : 50
Additional Details : Weight : 0.50000kg
Description
Human IL-17A Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL17 stimulates epithelial, endothelial or fibroblastic cells to secrete IL-6, IL-8 and G-CSF. IL17 also increases the expression of ICAM-1 in fibroblasts.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Cell Culture | |
31 kDa | |
25 ug | |
Human | |
Carrier-Free | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Neurobiology, Inflammation, Angiogenesis, Signal Transduction | |
98 % |
3605 | |
Sequential Chromatography | |
IL-17 | |
Store in freezer (-5 to -30°C). | |
25 μg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.