missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL13 Recombinant Mouse Protein

Recombinant Mouse Interleukin 13 (IL13) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMC0135

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10620744

  • £566.50 / Each


Estimated Shipment
08-02-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Disclaimers:

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.


Description

Description

IL13 down-regulates macrophage produced cytokines such as TNF-α, IL-1, IL-6, IL-8, and IL-12 in response to LPS challenge or IFN-γ. IL13 induces monocyte differentiation and also induces B cell differentiation and proliferation.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human TF-1 cells

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
16163
Sequential Chromatography
25μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
12kDa
98%
Mouse
Carrier Protein
Interleukins
IL-13
Cytokines & Receptors
Videos