Gibco™ IL12 Recombinant Human Protein

Recombinant IL12 is a bioactive protein intended for use in cell culture applications

Overview
Brand: Gibco™

Manufacturer Part Number: PHC1123

1 MG IL-12 RECOMBINANT HUMAN 1MG STORE AT 4°C

UNSPSC: 12352202

Code: 50

Additional Details:
Additional Details: Weight: 0.50000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 10226693

Quantity Price
1 £ 7070.0 / Each
Estimated Shipment date
from Supplier 31-01-2017
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description and Specification

Specification

Molecular Weight 75kDa
Purity 95%
Quantity 1mg
Source Human
Storage Requirements Store at -5° to -30°C.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype Interleukins
Endotoxin Concentration <0.1ng/μg
Gene Alias IL-12
Validated Application Cell Culture
Protein Family Cytokines & Receptors
Expression System Mammalian
Protein Length 503 aa
For Use With (Application) Cell Culture; Mammalian Cell Culture; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture
Format Lyophilized

IL12 induces the secretion of IFN-γ, IL-2 and TNF-α and stimulates the proliferation of PHA activated lymphocytes.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.