Learn More
Gibco™ Human IL-10 Recombinant Protein
Recombinant IL10 is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PHC0103
Code : 50
Additional Details : Weight : 0.56700kg
Description
Human IL-10 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL10 inhibits the synthesis of a number of cytokines, such as TNF-α, IL-1, and IL-6 from macrophages and IL-2, IFN-γ, and TNF-β from Th1 cells.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Cell Culture | |
Sequential Chromatography | |
IL-10 | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Signal Transduction, Stem Cell Research, Inflammation, Angiogenesis | |
95 % |
18.6 kDa | |
1mg | |
Human | |
Carrier-Free | |
1 mg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.