missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IGFBP6 Recombinant Human Protein

Recombinant Human Insulin-like Growth Factor Binding Protein 6 (IGFBP6) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC9621

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12033599

  • £896.10 / 100µg


Estimated Shipment
17-05-2018
Add to basket

Description

Description

IGFBP6 binds IGF-1 and II, prolonging the half life of these growth factors and altering the way these growth factors interact with cells surface receptors.
  • High purity: Minimizes interference from other proteins or contaminants
  • High biological activity: More results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: To help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to help ensure a biologically active protein
  • High Biological Activity: In-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by the dose dependent inhibition of IGF-II induced proliferation of MCF-7 cells

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
22.6kDa
100μg
Store at 2° to 8°C.
Recombinant/Ligand
<1 EU/mL
Inhibition of IGF-II induced proliferation of MCF-7 cells
Growth Factors & Receptors
0.4μg/mL
Lyophilized
95%
Human
Carrier Protein
IGFs (Insulin-Like Growth Factors)
IGFBP-6
Cell Culture
Mammalian
Videos

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.