Learn More
Gibco™ Human IGFBP-4 His-tag Recombinant Protein
Recombinant Human Insulin-like Growth Factor Binding Protein 4 (IGFBP4) is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PHC9614
Code : NEW
Additional Details : Weight : 1.00000kg
Description
Human IGFBP-4 His-tag Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IGFBP4 binds IGF1 and II, prolonging the half life of these growth factors and altering the way these growth factors interact with cells surface receptors.- High purity: minimizes interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to help ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by the dose dependent inhibition of IGF-II induced proliferation of MCF-7 cells
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
10μg | |
Human | |
Carrier-Free | |
inhibition of MCF-7 cell proliferation in the presence of 14 ng/mL IGF2 | |
10 μg | |
Mammalian | |
Recombinant, Ligand | |
IGFs (Insulin-Like Growth Factors) | |
Gibco™ |
26 kDa | |
IGFBP-4 | |
Store at 2 to 8°C | |
0.09 μg/ml | |
< 1 EU/ml | |
Wet Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Oncology, Apoptosis | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.