missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IGFBP4 Recombinant Human Protein

Recombinant Human Insulin-like Growth Factor Binding Protein 4 (IGFBP4) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC9611

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12013599

  • £799.28 / 100µg


Estimated Shipment
16-05-2018
Add to basket

Description

Description

IGFBP4 binds IGF1 and II, prolonging the half life of these growth factors and altering the way these growth factors interact with cells surface receptors.
  • High purity: minimizes interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to help ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by the dose dependent inhibition of IGF-II induced proliferation of MCF-7 cells

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
26kDa
100μg
Store at 2° to 8°C.
Recombinant/Ligand
<1 EU/mL
Inhibition of MCF-7 cell proliferation in the presence of 14ng/mL IGF2
Growth Factors & Receptors
0.09μg/mL
Lyophilized
95%
Human
Carrier Protein
IGFs (Insulin-Like Growth Factors)
IGFBP-4
Cell Culture
Mammalian
Videos

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.