missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IFN-γ Recombinant Rat Protein

Recombinant IFN-γ is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PRC4034

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10171013

  • £145.23 / Each


Estimated Shipment
10-07-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Recombinant rat IFN-γ is intended for use in cell culture applications. IFN-γ is involved in many biological processes such as inducing the secretion of TNF-α and stimulating the release of reactive oxygen species from macrophages. It is also involved with bone growth and inhibits bone reesorption. It is anti-viral and anti-parasitic and inhibits the proliferation of cells.

Shipping Condition: Dry Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Cell Signaling
25712
Sequential Chromatography
10μg
Store at -5° to -30°C.
Mutant/Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Liquid
16.9kDa
95%
Rat
Carrier Protein
Interferons
IFN-γ
Cytokines & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.