missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ GM-CSF Recombinant Swine Protein
Recombinant GM-CSF is a bioactive protein intended for use in cell culture applications
POA
Specifications
Purification Method | Sequential Chromatography |
---|---|
Gene Alias | GM-CSF |
Gene ID (Entrez) | 397208 |
Description
Recombinant swine GM-CSF is intended for use in cell culture applications. GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.Approved for shipment on Wet or Dry Ice.
Specifications
Sequential Chromatography | |
GM-CSF | |
397208 |
For Research Use Only. Not for use in diagnostic procedures.