Gibco™ GM-CSF Recombinant Swine Protein

Recombinant GM-CSF is a bioactive protein intended for use in cell culture applications

Overview
Brand: Gibco™

Manufacturer Part Number: PSC2015

UNSPSC: 12352202

Code: 50

Additional Details:
Additional Details: Weight: 0.50000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 10573775

Quantity Price
1 £ 276.0 / Each
Estimated Shipment date
from Supplier 10-04-2017
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description and Specification

Specification

For Use With (Application) Cell Culture; Mammalian Cell Culture
Gene ID (Entrez) 397208
Purification Method Sequential Chromatography
Protein Family Cytokines & Receptors
Endotoxin Concentration <0.1ng/μg
Quantity 25μg
Source Swine/Porcine
Purity 95%
Gene Alias GM-CSF
Expression System E. coli
Structural Form Recombinant/Ligand
Protein Subtype GM-CSF, G-CSF, & M-CSF
Validated Application Cell Culture
Storage Requirements Store at 2° to 8°C.
Molecular Weight 14kDa
Without Additives Carrier Protein
Format Lyophilized

Recombinant swine GM-CSF is intended for use in cell culture applications. GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.