missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ GM-CSF Recombinant Rat Protein

Recombinant GM-CSF is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PRC2011

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10058842

  • £1005.28 / Each


Estimated Shipment
12-03-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Recombinant rat GM-CSF is intended for use in cell culture applications. GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
116630
Exchange Chromatography
100μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
14.6kDa
95%
Rat
Carrier Protein
GM-CSF, G-CSF, & M-CSF
GM-CSF
Cytokines & Receptors
Videos