missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human GM-CSF, AOF Recombinant Protein
Human GM-CSF, AOF Recombinant Protein
Brand: Gibco™ PHC6025
Code : 50
Additional Details : Weight : 0.05400kg
Description
Human GM-CSF, AOF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human TF-1 cells
- Proven compatibility: Gibco proteins are bioassayed with Gibco media
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
20 ug | |
Human | |
Carrier-Free | |
proliferation of human TF-1 cells | |
20 μg | |
E. coli | |
Recombinant, Ligand | |
GM-CSF, G-CSF, & M-CSF | |
Gibco™ |
14.6 kDa | |
GM-CSF AOF | |
Store in freezer (-5 to -30°C). | |
0.1 ng/ml | |
< 1 EU/ml | |
Wet Ice | |
Cytokines & Receptors | |
Lyophilized | |
Signal Transduction, Stem Cell Research, Drug Development, Oncology | |
98 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.