missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ G-CSF Recombinant Mouse Protein

Recombinant G-CSF is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMC2035

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10496283

  • £286.34 / Each


Estimated Shipment
11-05-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

G-CSF stimulates the proliferation and differentiation of hematopoietic progenitor cells.

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Stem Cell Research; Stem Cell Culture; Stem Cell Differentiation; Hematopoietic Stem Cell Culture; Mesenchymal Stem Cell Culture; Differentiation & Development
12985
Sequential Chromatography
25μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
19kDa
95%
Mouse
Carrier Protein
GM-CSF, G-CSF, & M-CSF
G-CSF
Cytokines & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.