missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-9 Recombinant Protein
Human FGF-9 Recombinant Protein
Brand: Gibco™ PHG0194
Code : 50
Additional Details : Weight : 0.50000kg
Description
Human FGF-9 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
FGF9 (also known as heparin binding growth factor HBGF) is one of the isoforms of FGF-8, and is a member of the FGF family of proteins. It is involved in many biological processes including osteoblast proliferation and the survival of neurons.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Cell Culture | |
23.4 kDa | |
20 ug | |
Human | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Neurobiology, Oncology, Stem Cell Research, Microbiology, Differentiation, Angiogenesis | |
95 % |
2254 | |
Sequential Chromatography | |
FGF-9 | |
Store in freezer (-5 to -30°C). | |
20 μg | |
E. coli | |
Recombinant, Ligand | |
FGFs (Fibroblast Growth Factors) | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.