missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ FGF-8b Recombinant Human Protein

Recombinant Human Fibroblast Growth Factor 8B (FGF-8B) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHG0273

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10246693

  • £2147.55 / Each


Estimated Shipment
15-06-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

FGF-8b is one of the isoforms of FGF-8, and is involved in many biological processes including angiogenesis.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of BALB/3T3 cells

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Stem Cell Research; Stem Cell Culture; Stem Cell Differentiation; Embryonic Stem Cell Culture; Differentiation & Development
22.5kDa
95%
Human
Carrier Protein
FGFs (Fibroblast Growth Factors)
FGF-8b
Growth Factors & Receptors
Lyophilized
Sequential Chromatography
1mg
Store at 2° to 8°C.
Truncated/Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Videos

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.