Learn More
Gibco™ Embryonic Stem Cell FBS, Qualified, One Shot™ Format, US Origin
Secure sera for cell culture with least viral risk sera from BSE-free regions
Brand: Gibco™ 16141061
Code : 50
Additional Details : Weight : 0.10000kg
Description
- Sera Category: Specialty
- Origin: United States
- Specially tested for the ability to sustain undifferentiated cellular morphology of embryonic stem cells
- Endotoxin level: ≤50 EU/mL (levels routinely ≤ 10 EU/mL)
- Hemoglobin level: ≤25 mg/dL
Cell Culture, Embryonic Stem Cell Culture, Mammalian Cell Culture, Stem Cell Culture, Stem Cell Research
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Cattle/Bovine | |
Liquid | |
Fetal Bovine Serum | |
Standard (Sterile-filtered) | |
Sustainable packaging | |
Gibco™ |
Fetal | |
United States | |
Bottle | |
100 mL | |
Sterile | |
ES Cell-Qualified | |
Frozen |
For In Vitro Diagnostic Use.