missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Eotaxin Recombinant Human Protein
Recombinant human eotaxin is a bioactive protein intended for use in cell culture applications
POA
Specifications
Purification Method | Sequential Chromatography |
---|---|
Gene Alias | Eotaxin |
Gene ID (Entrez) | 6356 |
Description
Eotaxin is a member of the PF-4 family of chemokines. Eotaxin plays a role in inducing calcium flux in eosinophils.Shipping Condition: Wet Ice.
Specifications
Sequential Chromatography | |
Eotaxin | |
6356 |
For Research Use Only. Not for use in diagnostic procedures.