missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Eotaxin Recombinant Human Protein

Recombinant human eotaxin is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC1433

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10262543

  • £1888.80 / Each


Estimated Shipment
15-06-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Eotaxin is a member of the PF-4 family of chemokines. Eotaxin plays a role in inducing calcium flux in eosinophils.

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
6356
Sequential Chromatography
1mg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
∽8kDa
95%
Human
Carrier Protein
Eotaxins
Eotaxin
Chemokines & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.