missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CXCL1 (GROα) Recombinant Human Protein

Recombinant Human Chemokine (C-X-C motif) ligand 1 (Also known as CXCL1, GROα, melanoma growth stimulating activity, MSGA, Neutrophil-activating protein 3, NAP-3) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC1065

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10202873

  • £73.21 / Each


Estimated Shipment
15-06-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

CXCL1 is a chemokine that induces the infiltration of neutrophils.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by measuring the dose dependent mobilization of intracellular calcium (calcium flux) with human neutrophils
  • Proven Compatibility: Gibco proteins are bioassayed with Gibco media

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
2919
Sequential Chromatography
10μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
8kDa
95%
Human
Carrier Protein
GRO/KC/CINC Chemokines
CXCL1, GROα
Chemokines & Receptors
Videos

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.