missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Cripto Recombinant Human Protein

Recombinant human Cripto is a bioactive protein intended for use in cell culture applications.

Brand:  Gibco™ PHC8014

Code : 50

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10003664

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

Cripto is a member of the EGF-related family of growth factors.

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Stem Cell Research; Stem Cell Culture; Stem Cell Differentiation; Embryonic Stem Cell Culture; Hematopoietic Stem Cell Culture; Mesenchymal Stem Cell Culture; Differentiation & Development
4507424, 57932
Sequential Chromatography
10μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
Mammalian
Lyophilized
17.8kDa
95%
Human
Carrier Protein
EGF (Epidermal Growth Factor)
Cripto
Growth Factors & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.