missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ BMPR1A Recombinant Human Protein

Bone Morphogenetic Protein Receptor 1A (BMPR1A) Fc Chimera is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC9241

Code : 50

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 10216884

  • £471.74 / Each


Estimated Shipment
11-05-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

BMPR1A is one of the bone morphogenetic protein (BMP) receptors which are a family of transmembrane serine/threonine kinases that include the type I receptors BMPR1A and BMPR1B and the type II receptor BMPR2. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2. The ligands of these receptors are members of the TGF-beta superfamily. TGF-betas and activins transduce their signals through the formation of heteromeric complexes with 2 different types of serine (threonine) kinase receptors: type I receptors of about 50-55kDa and type II receptors of about 70-80kDa. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by its ability to inhibit human BMP4 induction of alkaline phosphatase in the mouse chondrogenic cell line ATDC5

Shipping Condition: Wet Ice.

Specifications

Specifications

NM_004329
Lyophilized
43.7kDa
95%
Human
Carrier Protein
BMPs (Bone Morphogenetic Proteins) & Related Factors
BMPR1A
Growth Factors & Receptors
ED50 range: 0.1 to 0.3μg/mL
Untagged
Cell Culture; Mammalian Cell Culture
BMPR1a
Sequential Chromatography
100μg
Store lyophilized human BMPR1a at 2° to 8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤ -20°C. Avoid repeated freeze/thaw cycles.
Recombinant/Fc Chimera/Ligand
<0.1ng/μg
Cell Culture
Mammalian
Human
Videos

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.