missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AmnioMAX™ C-100 Basal Medium

Part of kit used to prepare fully-supplemented Gibco AmnioMAX C-100 Complete Medium developed for short term culture of human amniotic fluid cells for cytogenetic studies and in vitro diagnostic procedures

Brand:  Gibco™ 17001074

Code : 50

Additional Details : Weight : 0.53000kg

 View more versions of this product

Product Code. 10744274

  • £136.00 / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Gibco AmnioMAX C-100 Complete Medium is two-part kit for preparation of a complete medium that requires no additional supplementation.

  • Quality and performance testing
  • Every lot of Gibco AmnioMAX C-100 Complete Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance
  • Pooled primary human amniotic fluid samples are cultured for six days in Gibco AmnioMAX C-100 Complete Medium before measuring total number of colonies and total number of mitotic colonies
  • In addition, each lot is tested for sterility, pH and osmolality
  • Unique, optimized formulation
  • Gibco AmnioMAX C-100 Complete Medium contains Fetal Bovine Serum (FBS), gentamicin, and L-glutamine to maximize cell attachment and growth
  • It has enhanced buffering system that provides greater pH stability during culture manipulations
  • Sustainable packaging

Amniotic Fluid and Chorionic Villus Culture, Cell Analysis, Cell Culture, Cell Proliferation, Cell Viability, Proliferation and Function, Clinical, Clinical and Translational Research, Cytogenetics, Mammalian Cell Culture

Specifications

Specifications

Storage conditions: 2 – 8°C
Shipping conditions: Ambient
Shelf life: 16 months from date of manufacture. Use complete medium within 7–10 days once opened.
Liquid
Standard Serum Supplementation
Glutamine, HEPES
AmnioMAX™
Room Temperature
Amniocyte Cell
Amniotic Fluid Cell Basal Medium
Sterile-filtered
Sustainable packaging
450 mL
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Intended Use: For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.