missing translation for 'onlineSavingsMsg'
Learn More

Gibberellic Acid A4+A7, 90% Alfa Aesar™

Brand:  Alfa Aesar J66661.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15428109

  • £154.20 / 5g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

Gibberellic Acid A4+A7

Specifications

Gibberellic Acid A4+A7
90%
582°C
Powder
Soluble in methanol, ethanol, or acetone; Also soluble in dilute solutions of sodium bicarbonate or sodium acetate
5g
Videos
SDS