missing translation for 'onlineSavingsMsg'
Learn More

Gibberellic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7059891
Click to view available options
Quantity:
500 mg
2 g
10 g
This item is not returnable. View return policy
77-06-5
C19H22O6
346.38
MFCD00079329
IXORZMNAPKEEDV-QTWFBFKQSA-N
gibberellic acid, gibberellin a3, gibberellin, gibreskol, brellin, cekugib, grocel, gibberellin x, gibberellic acid ga3, gib-tabs
91757643
(1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid
CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O
This item is not returnable. View return policy
77-06-5
C19H22O6
346.38
MFCD00079329
IXORZMNAPKEEDV-QTWFBFKQSA-N
gibberellic acid, gibberellin a3, gibberellin, gibreskol, brellin, cekugib, grocel, gibberellin x, gibberellic acid ga3, gib-tabs
91757643
(1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid
CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O

Plant growth hormone

Gibberellic acid is used as a plant growth hormone. It is also used to trigger germination in dormant seeds in laboratory and green house settings and to stimulate rapid stem and root growth and induce mitotic division in the leaves of some plants. It also serves in grape growing industry as a hormone to induce the production of larger bundles and bigger grape.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 222°C (decomposition)
Color White
Quantity 500 mg
Beilstein 54346
Merck Index 14,4419
Solubility Information Slightly soluble in water. Soluble in alcohol,isopropyl alcohol,ethanol,acetone and chloroform.
Optical Rotation +78° (c=2 in methanol)
Formula Weight 346.37
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Gibberellic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'