missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibberellic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mg
2 g
10 g
Description
Gibberellic acid is used as a plant growth hormone. It is also used to trigger germination in dormant seeds in laboratory and green house settings and to stimulate rapid stem and root growth and induce mitotic division in the leaves of some plants. It also serves in grape growing industry as a hormone to induce the production of larger bundles and bigger grape.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 222°C (decomposition) |
Color | White |
Quantity | 10 g |
Beilstein | 54346 |
Merck Index | 14,4419 |
Solubility Information | Slightly soluble in water. Soluble in alcohol,isopropyl alcohol,ethanol,acetone and chloroform. |
Optical Rotation | +78° (c=2 in methanol) |
Formula Weight | 346.37 |
Percent Purity | 98% |
Physical Form | Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'