missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gibberellic acid, 98%

Plant growth hormone

Brand:  Alfa Aesar™ 045569.ME

Code : NEW

Additional Details : CAS Number : 77-06-5 Weight : 0.00050kg

Quantity 500mg

 View more versions of this product

Product Code. 15402377

  • £20.50 / 500mg

Estimated Shipment: 04-10-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

77-06-5
346.38
IXORZMNAPKEEDV-QTWFBFKQSA-N
91757643
CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O
C19H22O6
MFCD00079329
gibberellic acid, gibberellin a3, gibberellin, gibreskol, brellin, cekugib, grocel, gibberellin x, gibberellic acid ga3, gib-tabs
(1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid

Specifications

Gibberellic acid
White
500mg
C19H22O6
54346
gibberellic acid, gibberellin a3, gibberellin, gibreskol, brellin, cekugib, grocel, gibberellin x, gibberellic acid ga3, gib-tabs
IXORZMNAPKEEDV-QTWFBFKQSA-N
(1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid
91757643
Powder
+78° (c=2 in methanol)
222°C (decomposition)
77-06-5
MFCD00079329
14,4419
Slightly soluble in water. Soluble in alcohol,isopropyl alcohol,ethanol,acetone and chloroform.
CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O
346.38
346.37
98%
Videos
SDS Documents
Certificates