missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Giant Stir Bar

Designed for use in large containers and vials. These giant PTFE-coated magnetic stir bares are autoclavable.

Brand:  Fisherbrand™

Code : 41

Additional Details : Weight : 0.03008kg

 View more versions of this product

Product Code. 10480971

  • £8.33 / Each


In Stock 24
Add to basket

You may only order up to the available stock. For more details call customer services on 01509 555 500

Description

Description

  • Stable, general-purpose magnetic stir bar
  • Designed for use in large containers and vials
  • Autoclavable
Specifications

Specifications

Yes
PTFE
PTFE encapsulated Alnico V magnet core
2/Pk.
14mm
Triangular
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
White
Containers & Vessels / Stirrer Hot Plates
Giant PTFE Stir Bar
80mm
Videos