missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ GFRA1 Recombinant Rat Protein, His Tag

Expressed from DNA sequence encoding rat GFRA1 (Met 1-Leu 445) fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 80021R08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15339506

  • £123.88 / 25µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
25454
≥90% by SDS-PAGE
Rat
<1 EU/μg
GFRA1
Growth Factors and Receptors
GFRA1 Recombinant Rat Protein
Lyophilized
48kd
5 x 5μg
Full Length, Recombinant
GDNF Family Receptor Alpha 1
Assay Standard
Mammalian
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.