missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GFR alpha-1/GDNF R alpha-1 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | GFR alpha-1/GDNF R alpha-1 |
---|---|
Clone | 260714 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
GFR alpha-1/GDNF R alpha-1 Monoclonal specifically detects GFR alpha-1/GDNF R alpha-1 in Human samples. It is validated for Western Blot.Specifications
GFR alpha-1/GDNF R alpha-1 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P56159 | |
2674 | |
Mouse myeloma cell line NS0-derived recombinant human GFR alpha-1 Asp25-Lys434 Accession # P56159 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human GFR alpha-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human GFR alpha-2, -3, -4, or recombinant rat GFR alpha-1 is observed. |
260714 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp313E1012, DKFZp686J0156, GDNF family receptor alpha 1, GDNF family receptor alpha-1, GDNF R alpha-1, GDNF receptor alpha-1, GDNFRAGFR-ALPHA-1, GDNFR-alpha-1, GDNFRMGC23045, GFR alpha1, GFRa-1, GFRA1, GFR-alpha-1, Glial cell line-derived neurotrophic factor receptor alpha, GPI-linked anchor protein, PI-linked cell-surface accessory protein, RET ligand 1, RET1L, RETL1FLJ10561, TGF-beta related neurotrophic factor receptor 1, TGF-beta-related neurotrophic factor receptor 1, TRNR1FLJ31546 | |
GFRA1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only