missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | GFP |
---|---|
Clone | 454518 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
GFP Monoclonal specifically detects GFP in Multi-Species samples. It is validated for Western Blot, Immunoprecipitation.Specifications
GFP | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Multi-species | |
P42212 | |
GFP | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
454518 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
eGFP, Enhanced Green Fluorescent Protein, GFPuv, Green Fluorescent Protein, green fluorescent protein (gfp) | |
E. coli-derived recombinant GFPuv Ala2-Tyr238 Accession # P42212 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects GFPuv in direct ELISAs and Western blots. |
For Research Use Only