missing translation for 'onlineSavingsMsg'
Learn More

GFP Rat anti-Tag, eFluor® 660, Clone: 5F12.4, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-6498-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15589996

  • £177.00 / 25µg

Estimated Shipment
02-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. The jellyfish Aequorea victoria contains green fluorescent protein (GFP) that emits light in the bioluminescence reaction of the animal. GFP has been used widely as a reporter protein for gene expression in eukaryotic and prokaryotic organinsms, and as a protein tag in cell culture and in multicellular organisms. As a fusion tag, GFP can be used to localize proteins, to study their movement or to research the dynamics of the subcelluclar compartments where these proteins are targeted. GFP technology has revealed considerable new insights in the physiological activities of living cells. GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species independent and is suitable for a variety of applications. GFP has been used etensively as a fluorescent tag to monitor gene expressin and protein localization. Moreover, other applications for GFP include its use in assessing protein protein interactions in the yeast two hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.
Specifications

Specifications

GFP
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
GFP2; Green Fluoresecent Protein; GFP; Turbo GFP; eGFP
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
Tag
Flow Cytometry
5F12.4
eFluor™ 660
Liquid
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Videos
Documents

For Research Use Only.