missing translation for 'onlineSavingsMsg'
Learn More

Germanium Standard For ICP/MS, SPEX CertiPrep™

Catalog Number p-218331
Click to view available options
Quantity:
30 mL
125 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
7732-18-5
This item is not returnable. View return policy
7732-18-5

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ge10μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Grade Trace ICP/ICP-MS
Packaging Triple-leached LDPE bottle
Solvent or Matrix H2O/Tr. F-
Quantity 30 mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Chemical Name or Material Germanium Standard

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'