missing translation for 'onlineSavingsMsg'
Learn More

Germanium rod, polycrystalline, 6.35mm (0.25in) dia, 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog Number 15464885
Click to view available options
Quantity:
10 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 7440-56-4
Molecular Formula Ge
MDL Number MFCD00085310
Quantity 10 cm
Synonym element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
InChI Key GNPVGFCGXDBREM-UHFFFAOYSA-N
SMILES [Ge]
IUPAC Name germanium
Molecular Weight (g/mol) 72.63
PubChem CID 6326954
ChEBI CHEBI:30441
Percent Purity 99.9999%
Odor Odorless
Weight ≈1.7g/cm
Form Polycrystalline
Assay (metals basis)
Chemical Name or Material Germanium rod, 6.35mm (0.25 in.) dia.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'