missing translation for 'onlineSavingsMsg'
Learn More

Germanium, plasma standard solution, Specpure™ Ge 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-7051914
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
MFCD00085310
This item is not returnable. View return policy
MFCD00085310

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 mL
Grade Specpure™
Physical Form Liquid
Chemical Name or Material Germanium plasma standard solution, Ge 1000μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'