missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Germanium, plasma standard solution, Specpure™, Ge 10,000μg/mL

Brand:  Alfa Aesar™ 014410.AE

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 3264 Chem class : 8 Pack group : III

Quantity 100mL

 View more versions of this product

Product Code. 11302348

  • £91.30 / 100mL


Estimated Shipment date
from Supplier 21-06-2018
Add to basket

Specifications

Chemical Identifiers

Germanium plasma standard solution
Specpure™
UN3264
Ge 10000μg/mL
MFCD00085310

Specifications

Germanium plasma standard solution
Specpure™
Ge in 5% HNO3/tr. HF
UN3264
Ge 10000μg/mL
Liquid
MFCD00085310
100mL
Videos
SDS