missing translation for 'onlineSavingsMsg'
Learn More

Germanium pieces, 2cm (0.8in) & down, 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048195
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
7440-56-4
Ge
72.63
MFCD00085310
GNPVGFCGXDBREM-UHFFFAOYSA-N
element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
6326954
CHEBI:30441
germanium
[Ge]
This item is not returnable. View return policy
7440-56-4
Ge
72.63
MFCD00085310
GNPVGFCGXDBREM-UHFFFAOYSA-N
element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
6326954
CHEBI:30441
germanium
[Ge]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 10 g
Percent Purity 99.9999%
Odor Odorless
Particle Size 2cm (0.8 in.) & down
Form Pieces
Assay (metals basis)
Chemical Name or Material Germanium pieces, Electrical Resistivity: 40 ohm-cm

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'