missing translation for 'onlineSavingsMsg'
Learn More

Germanium n-butoxide, 97+%, Thermo Scientific Chemicals

Catalog Number p-7051229
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
25063-27-8
MFCD00015347
Tetra-n-butoxygermane
This item is not returnable. View return policy
25063-27-8
MFCD00015347
Tetra-n-butoxygermane

Oxide films

Germanium n-butoxide is used as oxide films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.017 g/mL
Boiling Point 143°C (8 mmHg)
Flash Point 112°C (234°F)
Linear Formula Ge[O(CH2)3CH3]4
Refractive Index 1.4255
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 365.05
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Germanium n-butoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'