missing translation for 'onlineSavingsMsg'
Learn More

Germanium(IV) ethoxide, 97%, Thermo Scientific Chemicals

Catalog Number p-7049553
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
14165-55-0
MFCD00015121
Germanium tetraethoxide; Tetraethoxygermane
This item is not returnable. View return policy
14165-55-0
MFCD00015121
Germanium tetraethoxide; Tetraethoxygermane

It is a sol-gel intermediate for optical fibers and photosensitive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -72°C
Color Colorless
Density 1.14 g/mL
Boiling Point 185.5°C
Flash Point 49°C (121°F)
Odor Alcohol-like
Linear Formula Ge(OC2H5)4
Refractive Index 1.407
Quantity 1 g
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Soluble in anhydrous alcohol and benzene; Hydrolyzes in water
Formula Weight 252.84
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Germanium(IV) ethoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'