missing translation for 'onlineSavingsMsg'
Learn More

Germanium(IV) chloride, 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048428
Click to view available options
Quantity:
0.5 g
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
10038-98-9
Cl4GeH8
222.49
MFCD00011029
VDHBHZZBEGQJRK-UHFFFAOYSA-N
germanium tetrachloride, germanium chloride, germane, tetrachloro, germanium iv chloride, germanium, tetrachloro, gecl4, germanium chloride gecl4, unii-ysv1r803c0, wln: .ge..g4, germanium chloride gecl4 8ci
66226
Cl.Cl.Cl.Cl.[GeH4]
This item is not returnable. View return policy
10038-98-9
Cl4GeH8
222.49
MFCD00011029
VDHBHZZBEGQJRK-UHFFFAOYSA-N
germanium tetrachloride, germanium chloride, germane, tetrachloro, germanium iv chloride, germanium, tetrachloro, gecl4, germanium chloride gecl4, unii-ysv1r803c0, wln: .ge..g4, germanium chloride gecl4 8ci
66226
Cl.Cl.Cl.Cl.[GeH4]

It is used as a catalyst in the conversion of carbohydrates into 5-hydroxymethylfurfural in ionic liquids, as a reducing agent in combination with triphenylphosphine (TPP) for the reduction of alpha-bromo carboxylic acid derivatives, and in the production of pure germanium. It also finds use as an intermediate for several optical processes. It is one of the most important dopants in silica glass for optical fibers. It is employed in the preparation of germanium dioxide which is used for wide camera lens, microscopy, IR-transparent glasses/windows/lenses, and for the core of fiber-optic lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 83.1°C
Melting Point -49.5°C
Color Colorless
Physical Form Liquid
Quantity 0.5 g
Linear Formula GeCl4
UN Number UN3264
Merck Index 14,4410
IUPAC Name germanium tetrachloride
Formula Weight 214.4
Percent Purity 99.9999%
Odor Acidic
Sensitivity Moisture Sensitive
Density 1.874g/mL
Assay (metals basis)
Chemical Name or Material Germanium(IV) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'