Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Simplify sampling with easy access
Brand:  Thermo Scientific™ 840-277700
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15540592
  • £215.00 / Each
Estimated Shipment: 29-05-2024
to see stock.
Description
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.
Specifications
Specifications
Long path rectangular cell holder that accommodates 20-100 mm path length.  Comes assembled in a removable compartment liner.
Measurements of solutions with low absorbance values
20 to 100 mm cuvette (1)
Long Path Cell Holder
GENESYS 30/40/50 Spectrophotometers
Rectangular