missing translation for 'onlineSavingsMsg'
Learn More

Brand™ General Purpose Hydrometer

Brand:  Brand™ 966037

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 10439451

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

1Pack
1.000 to 1.060g/cm3
General Purpose Hydrometer
160mm
0.001g/cm3
Videos