missing translation for 'onlineSavingsMsg'
Learn More

General-Purpose BioPharm Silicone Tubing for FH100 and FH100X Pumps

General-Purpose BioPharm Silicone Tubing is suitable for most everyday applications where pressure, accuracy, and long-term pump life are not critical to the process.

Brand:  Thermo Scientific Barnant 72-300-017

Code : Z4

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12706767

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

BioPharm Silicone Tubing (platinum-cured)

  • Tested to operate in Thermo Scientific peristaltic pumps
  • Ultra-smooth inner surface minimizes particle entrapment
  • Very low extractables with documented biocompatibility for sensitive applications
  • Ideal for lab, biotech and pharmaceutical applications
  • Supplied 15.2m (50ft.) per pack
Specifications

Specifications

USP Class VI. FDA 21 CFR 177.2600, Exceeds 3A sanitary standards, European Pharmacopoeia (EP)
Translucent, clear to light amber
Thermal-set Rubber, Siloxane Polymers, Amorphous Silica
Ultra-smooth inner surface minimizes particle entrapment. Lower absorption; excellent biocompatibility; no leachable additive, DOP, or plasticizers. Very low extractables. Odorless and nontoxic, fungus-resistant. No taste imparted to transported fluids. Weather, ozone, corona, and radiation resistant.
FH100
Do not use with concentrated solvents, oils, acids. Relatively high gas permeability.
-60°C
Sterilize by ETO, autoclave, or gamma radiation up to 2.5 Mrad. To autoclave: coil loosely in nonlinting cloth or paper; autoclave at 121°C (250°F), 1 bar (15 psi) for 30 minutes.
17
Good
6.4mm
24 to 2000mL/min.
1520cm
BioPharm Silicone
CO2: 25,147; H2: ----; O2: 4715; N2: 2284
232°C
50 ft.
1.6mm
Good
Videos
Documentation