Thermo Scientific™ GeneJET™ Plasmid Miniprep Kit PROMO
Perform isolation of high quality plasmid DNA from recombinant E. coli cultures with this simple, rapid, and cost-effective system.
Overview
Brand: Thermo Scientific™
Manufacturer Part Number: K0503-RMP6225
70ML GeneJET Plasmid Miniprep Kit
UNSPSC: 41105504
Additional Details:
Additional Details: Weight: 1.35000kg Transport: UN number : 1824 Chem class : 1b Pack group : III
Product Code. 15573561
Quantity Price
1 £ 205.0 / 70mL
Estimated Shipment date
from Supplier 30-01-2017
Add to basket
Description and Specification
Highlights
  • Efficient -high yields of up to 20 µg of high quality plasmid DNA
  • Fast -procedure takes less than 14 minutes
  • Convenient -no phenol/chloroform extraction or alcohol precipitation required
  • Pure -purified DNA is immediately ready to use