Thermo Scientific™ GeneJET™ Plasmid Miniprep Kit PROMO
Perform isolation of high quality plasmid DNA from recombinant E. coli cultures with this simple, rapid, and cost-effective system.
Overview
Brand: Thermo Scientific™
Manufacturer Part Number: K0502-RMP6225
15ML GeneJET Plasmid Miniprep Kit
UNSPSC: 41105504
Additional Details:
Additional Details: Weight: 0.10000kg Transport: UN number : 1824 Chem class : 1b Pack group : III
Product Code. 15563561
Quantity Price
1 £ 49.78 / 75mL
EU Stock 119
Add to basket
Description and Specification
Highlights
  • Efficient -high yields of up to 20 µg of high quality plasmid DNA
  • Fast -procedure takes less than 14 minutes
  • Convenient -no phenol/chloroform extraction or alcohol precipitation required
  • Pure -purified DNA is immediately ready to use